Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AATB-13410

AAT Bioquest

AAT Bioquest Ac-IETD-AFC

5 mg

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$232.00

$232.00


Available to ship onApril 22, 2025Details



Select a Size

Change View
1 EA
$232.00

About This Item

CAS Number:
Molecular Weight:
729.65
PDF Icon
Shipped & Sold By AAT Bioquest
A Trusted Marketplace Seller
PDF Icon
Have a question?

$232.00


Available to ship onApril 22, 2025Details


fluorescence

λex 376 nm;λem 482 nm

manufacturer/tradename

AAT Bioquest

General description

Ac-IETD-AFC is a fluorogenic caspase-8/granzyme B substrate containing the acetyl (Ac) moiety.

Ac-IETD-AFC *CAS 211990-57-7* is manufactured by AAT Bioquest

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library