Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AABH97D2405A

AA BLOCKS, INC.

ethyl 2-cyano-3-[3-(trifluoromethoxy)phenyl]prop-2-enoate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
285.22
MDL number:

Iupac Name

ethyl (E)-2-cyano-3-(3-(trifluoromethoxy)phenyl)acrylate

Inchi Code

1S/C13H10F3NO3/c1-2-19-12(18)10(8-17)6-9-4-3-5-11(7-9)20-13(14,15)16/h3-7H,2H2,1H3/b10-6+

InChI key

AZHDVDXWRLOFJB-UXBLZVDNSA-N

storage temp.

2~8 C

Purity

97%

Country of Origin

US

Product Link

https://www.aablocks.com/prod/773089-44-4

Synonym

ethyl 2-cyano-3-(3-(trifluoromethoxy)phenyl)acrylate

MSDS

https://www.aablocks.com/downloadSds?cas=773089-44-4


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library