AABH3036E3E0
AA BLOCKS, INC.
2-(2,3-dihydro-1H-indol-1-yl)acetic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
2,3-dihydro-1H-indol-1-ylacetic acid
Inchi Code
1S/C10H11NO2/c12-10(13)7-11-6-5-8-3-1-2-4-9(8)11/h1-4H,5-7H2,(H,12,13)
InChI key
ZYKYVZPUUGVMFE-UHFFFAOYSA-N
storage temp.
2~8 C
Country of Origin
US
Product Link
https://www.aablocks.com/prod/193544-62-6
Synonym
2,3-dihydro-1H-indol-1-ylacetic acid
MSDS
https://www.aablocks.com/downloadSds?cas=193544-62-6
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.