SRP6164
G-CSF human
recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)
Synonym(s):
CSF-3, Filgrastim, GM-CSF beta, MGI-1G, Pluripoietin
About This Item
Recommended Products
biological source
human
recombinant
expressed in HEK 293 cells
assay
≥95% (SDS-PAGE)
form
lyophilized
potency
0.02-0.08 ng/mL
mol wt
21-25 kDa (monomer, glycosylated)
packaging
pkg of 10 μg
storage condition
avoid repeated freeze/thaw cycles
technique(s)
cell culture | mammalian: suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
Gene Information
human ... CSF3(1440)
General description
Biochem/physiol Actions
Physical form
Preparation Note
Reconstitution
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service