Skip to Content
MilliporeSigma
All Photos(1)

Documents

MAK414

Sigma-Aldrich

COX Activity Assay Kit (Fluorometric)

sufficient for 100 fluorometric tests

Synonym(s):

Cyclooxygenase Activity Assay Kit (Fluorometric)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503

detection method

fluorometric

relevant disease(s)

rheumatological diseases

storage temp.

−20°C

General description

Cyclooxygenase (COX), also known as prostaglandin-endoperoxide synthase (PTGS), is an enzyme that is responsible for the formation of important biological mediators called prostanoids, including prostaglandins, prostacyclin and thromboxane. COX is the central enzyme in the biosynthetic pathway to prostanoids from arachidonic acid. There are two known isoenzymes: COX-1 and COX-2. COX-1 is constitutively expressed in many tissues and is the predominant form in gastric mucosa and in kidney. COX-2 is not expressed under normal conditions in most cells, but elevated levels are observed during inflammation. Pharmacological inhibition of COX by non-steroidal anti-inflammatory drugs (NSAID) can provide relief from the symptoms of inflammation and pain.

Features and Benefits

COX Activity Assay Kit:
  • Compatible with high-throughput handling systems.
  • Includes COX-1 inhibitor (SC560) & COX-2 Inhibitor (Celecoxib).

Suitability

The kit is suitable for the measurement of COX activity in various biological samples (such as adherent and suspension cells and tissues) and purified/crude enzyme preparations.

Principle

The COX Activity Assay Kit provides a simple method to detect the peroxidase activity of COX in biological samples or purified/crude enzyme preparations. The kit includes COX-1 and COX-2 specific inhibitors to differentiate the activity of COX-1 and COX-2, as well as other peroxidases, which may be present in the sample.

Storage Class

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service