Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

27102

Supelco

Chromoionophore XI

Selectophore

Synonym(s):

ETH 7061, Fluorescein octadecyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H48O5
CAS Number:
Molecular Weight:
584.78
Beilstein/REAXYS Number:
6626911
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

product line

Selectophore

form

powder

SMILES string

CCCCCCCCCCCCCCCCCCOC(=O)c1ccccc1C2=C3C=CC(=O)C=C3Oc4cc(O)ccc24

InChI

1S/C38H48O5/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-19-26-42-38(41)32-21-18-17-20-31(32)37-33-24-22-29(39)27-35(33)43-36-28-30(40)23-25-34(36)37/h17-18,20-25,27-28,39H,2-16,19,26H2,1H3

InChI key

UDHKRSZNYLYOEP-UHFFFAOYSA-N

General description

Chromoionophore XI (fluorescein octadecyl ester, ETH 7061) is a lipophilized pH indicator.[1]
Visit our Sensor Applications portal to learn more.

Application

Selected application examples
ETH 7061 may be used in bulk flow optode, used for determination of perchlorate.[1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Chromoionophore in chloride-selective optodes[2]; Ionophore used in an alcohol-selective electrode[3]; Lipophilic pH indicator[4]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H H Zeng et al.
Talanta, 41(6), 969-975 (1994-06-01)
A new type of optode membrane for the determination of alcohols, such as methanol, ethanol, propan-1-ol and propan-2-ol etc, is presented that can be exploited in optical and fibre-optic sensors. It is based on the use of a new lipophilic
L. Brown et al.
Journal of the Chemical Society. Perkin Transactions 1, 3349-3349 (1990)
Flow-through bulk optode for the fluorimetric determination of perchlorate.
Ortuna JA
Talanta, 60 (2), 563-569 (2003)
S.S. Tan et al.
Analytica Chimica Acta, 255, 35-35 (1991)
Weihai Zhang et al.
Acta biomaterialia, 116, 344-355 (2020-09-02)
The uptake and trafficking of NPs is impacted by several attributes such as size, shape, surface charge and importantly by surface ligands that can interact with the cell plasma membrane. We envision that NPs which can be readily modified in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service