Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

754439

Sigma-Aldrich

4-Nitrobenzylphosphonic acid

97%

Synonym(s):

p-Nitrobenzylphosphonic acid, [(4-Nitrophenyl)methyl]phosphonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8NO5P
CAS Number:
Molecular Weight:
217.12
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

97%

form

solid

mp

222-228 °C

SMILES string

OP(O)(=O)Cc1ccc(cc1)[N+]([O-])=O

InChI

1S/C7H8NO5P/c9-8(10)7-3-1-6(2-4-7)5-14(11,12)13/h1-4H,5H2,(H2,11,12,13)

InChI key

FZNXRFYRXBFQMX-UHFFFAOYSA-N

Application

This molecule is capable of self-assembling on a variety of oxide surfaces. It has also been used to tune the electronic properties of electrodes in Dye-Sensitized Solar Cell Devices.[1]

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sven Rühle et al.
The journal of physical chemistry. B, 109(40), 18907-18913 (2006-07-21)
Molecular modification of dye-sensitized, mesoporous TiO2 electrodes changes their electronic properties. We show that the open-circuit voltage (V(oc)) of dye-sensitized solar cells varies linearly with the dipole moment of coadsorbed phosphonic, benzoic, and dicarboxylic acid derivatives. A similar dependence is

Articles

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service