Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

579505

Sigma-Aldrich

3-(Trifluoromethoxy)benzenesulfonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3OC6H4SO2Cl
CAS Number:
Molecular Weight:
260.62
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

refractive index

n20/D 1.4760 (lit.)

bp

229-230 °C (lit.)

density

1.530 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)Oc1cccc(c1)S(Cl)(=O)=O

InChI

1S/C7H4ClF3O3S/c8-15(12,13)6-3-1-2-5(4-6)14-7(9,10)11/h1-4H

InChI key

DODDSXTWDSJCDN-UHFFFAOYSA-N

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

>230.0 °F - closed cup

flash_point_c

> 110 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service