跳轉至內容
Merck
  • Conformationally constrained mimetics of laminin peptide YIGSR as precursors for antimetastatic disintegrins.
Conformationally constrained mimetics of laminin peptide YIGSR as precursors for antimetastatic disintegrins.
Journal of medicinal chemistry (2009-12-03)
Angelo Bella, Helen Lewis, Jennifer Phu, Andrew R Bottrill, Sharad C Mistry, Christine E Pullar, Maxim G Ryadnov
摘要
Conformationally constrained mimetics of the laminin cell-adhesion site, YIGSR, are described. The site is the natural antagonist of the integrin-associated laminin receptor 1 (LAMR1) known to mediate metastatic tumor adhesion. The attachment of selected metastatic cell lines toward the constrained antagonists has been assessed. Observed differential responses prompted by folding preferences of the mimetics revealed stronger attachment activities for turnlike structures. The results permit the conformational design of antimetastatic disintegrins.
材料
產品編號
品牌
產品描述
Sigma-Aldrich
Tyr-Ile-Gly-Ser-Arg, ≥97% (HPLC)