文件
509515
SGE 旋转锁定气密注射器
25MDR-LL-GT, volume 25 mL, needle size (not included)
同義詞:
SGE 25mL 旋转锁定注射器
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41122001
材料
glass syringe
針尺寸
(not included)
特點
needle type Luer lock
包裝
pkg of 1 ea
製造商/商標名
SGE 009462
容積
25 mL
尋找類似的產品? 前往 產品比較指南
一般說明
  • SGE gas tight syringe without a needle, Model: 25MDR-LL-GT.
  • Made of borosilicate glass, PTFE plunger tips, and KEL-F Luer cones.
  • Autoclavable syringes but the plunger should be removed.
  • Features an interference fit between the plunger tip and the barrel which enhances the gas sealing properties of the syringe.
  • Maximum inertness for liquid or gas samples.
Choose from one of the most recent versions:
分析證明 (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact 客戶支援