24407
1 of 10
24523
Hamilton® 注射器,用于 PerkinElmer® GC Autosystem 和 Clarus 500
21929-U
用于 PerkinElmer® Autosystem 气相色谱仪的 SGE 注射器
HAM80075
Hamilton® syringe
21521
SGE 导向推杆注射器
509272
SGE 微量注射器
24400
Supelco
5.33002
甲酸98%-100%
47650-U
CARB 方法 1004 DNPH 混标 1
CADG5177
Dissolved gas analysis (DGA) glass syringe
24408
cone tip stainless steel needle (point style 1)
23 ga
color (light orange)liquid syringe
removable needle
pkg of 1 ea
SGE 000478
70 mm
0.5 μL
尋找類似的產品? 前往 產品比較指南
默克Supelco®分析化学_分析色谱_分析进样针-默克生命科学
Choose from one of the most recent versions:
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact 客戶支援