文件
Z329789
BRAND® wash bottle, wide-mouth, labelled, LDPE
capacity 500 mL, label, Acetone, without venting valve
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41103209
NACRES:
NB.17
材料
low-density polyethylene bottle
red closure
round bottle
translucent bottle
wide-mouth bottle
特點
label Acetone
with cap
without venting valve
包裝
pack of 5 ea
製造商/商標名
BRAND 144085
容量
500 mL
尋找類似的產品? 前往 產品比較指南
一般說明
Flexible, high-quality wash bottles with integral wash steam and sealing cone in the closure. Large opening for convenient filling. Label with designation of contents and general safety statements (with CAS no. and NFPA codes).
Bottles with venting valve (metal ball) are for distilled water and organic solvents only; to avoid dropping caused by overpressure.
其他說明
BR144085 (India)
法律資訊
BRAND is a registered trademark of BRAND GMBH + CO KG
附屬品