化驗
≥95% (HPLC)
成份
Peptide content, ~65%
儲存溫度
−20°C
SMILES 字串
CC(C)CC(NC(=O)C(Cc1cnc[nH]1)NC(=O)C(Cc2ccccc2)NC(=O)C3CCCN3C(=O)C(N)Cc4cnc[nH]4)C(=O)NC(CC(C)C)C(=O)NC(C(C)C)C(=O)NC(Cc5ccc(O)cc5)C(O)=O
生化/生理作用
Potent inhibitor of renin at acidic pH. Can be linked to agarose and used to purify renin, which elutes at neutral pH.
儲存類別代碼
13 - Non Combustible Solids
水污染物質分類(WGK)
WGK 3
閃點(°F)
Not applicable
閃點(°C)
Not applicable
個人防護裝備
Eyeshields, Gloves, type N95 (US)
E E Slater et al.
The Journal of biological chemistry, 256(15), 8164-8171 (1981-08-10)
Human renal renin was purified from normal kidney by either of two protocols which combined sequential DEAE-cellulose chromatography, pepstatin affinity chromatography, gel filtration, and a final step of affinity chromatography using either the synthetic octapeptide renin inhibitor (D-Leu6] or antirenin
Competitive inhibitors of renin.
K Poulsen et al.
Biochemistry, 12(20), 3877-3882 (1973-09-25)
我們的科學家團隊在所有研究領域都有豐富的經驗,包括生命科學、材料科學、化學合成、色譜、分析等.
聯絡技術服務