重要文件
EPM1289-0500
Test Tube Rack, for Eppendorf® New Brunswick Innova® and Eppendorf® Excella® Shakers
size large, holds 22 tubes
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41122804
NACRES:
NB.73
尺寸
large
450x80x129 mm
特點
holds 22 tubes
製造商/商標名
Eppendorf® M1289-0500 (Global)
尋找類似的產品? 前往 產品比較指南
一般說明
Test Tube Racks have an adjustable angle and tube height ­support. Made from stainless-steel with inserts to help retain tubes and ­prevent them from spinning in the rack. Finger-tight knobs let you quickly adjust angle, or release the rack from the base plate, for easy transport to the bench.
法律資訊
Eppendorf is a registered trademark of Eppendorf AG
Excella is a registered trademark of Eppendorf, Inc. 
Innova is a registered trademark of Eppendorf, Inc. 
New Brunswick is a trademark of Eppendorf AG
從最近期的版本中選擇一個:
分析證明 (COA)