重要文件
EP951020594
Eppendorf® twin.tec PCR Plates 384
Optimized rim for barcodes, PCR clean, colorless, pack of 300 ea
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41106300
材料
colorless
conical bottom
polypropylene wells
無菌
non-sterile
特點
PCR clean
lid: no
skirt
包裝
pack of 300 ea
製造商/商標名
Eppendorf® 951020594
尺寸
384 well
孔容積
45 μL
適合性
suitable for PCR
尋找類似的產品? 前往 產品比較指南
相關類別
一般說明
  • Eppendorf® twin.tec PCR Plate 384, 45 µL, PCR clean, colorless, 300 plates
  • Optimized rim for barcodes and automated work processes
  • Stackable
  • Within SBS footprint recommendations
  • Ideal for most common 384-well thermocyclers, particularly for Eppendorf Mastercycler pro 384
  • 40 µL max. well volume
  • Available with barcode (upon request), for skirted and semi-skirted only
Eppendorf® twin.tec PCR Plate 384, 45 µL, PCR clean, red, 300 plates
  • Optimized rim for barcodes and automated work processes
  • Stackable
  • Within SBS footprint recommendations
  • Ideal for most common 384-well thermocyclers, particularly for Eppendorf Mastercycler pro 384
  • 40 µL max. well volume
特點和優勢
  • Optimized rim for barcodes and automated work processes
  • Stackable
  • Within SBS footprint recommendations
  • Ideal for most common 384-well thermocyclers, particularly for Eppendorf Mastercycler pro 384
  • 40 µL max. well volume
  • Available with barcode (upon request), for skirted and semi-skirted only
法律資訊
Eppendorf is a registered trademark of Eppendorf AG
從最近期的版本中選擇一個:
分析證明 (COA)