重要文件
EP5405000174
Eppendorf® Pharma Package Centrifuge 5425 G
refrigerated: no, with rotary knobs, without standard rotor, with FA-18x2 rotor kit, AC/DC input 230 V, 50 - 60 Hz, EuroPlug
登入查看組織和合約定價
About This Item
分類程式碼代碼:
40161701
NACRES:
NB.81
AC/DC 輸入
230 V, 50 - 60 Hz, EuroPlug
特點
CE compliant
CSA approved
refrigerated: no
包裝
pack of 1 ea
製造商/商標名
Eppendorf® 5405000174
尋找類似的產品? 前往 產品比較指南
成分
With rotary knobs, without standard rotor, with FA-18x2 rotor kit
其他說明
This rotor is not compatible with the first generation of centrifuge 5804/5804R up to serial number 10000 and 5810R from before serial number 12000.
法律資訊
Eppendorf is a registered trademark of Eppendorf AG
從最近期的版本中選擇一個:
分析證明 (COA)