sterility
sterile
manufacturer/tradename
Corning 3003
L
247 mm
General description
康宁一次性刮刀旨在节省时间并提供无污染的样品。减少了使用可重复使用的刮刀的回收和消毒步骤。
每个刮刀都是独立包装的,不含 RNase/DNase、无热原和抗静电。
每个刮刀都是独立包装的,不含 RNase/DNase、无热原和抗静电。
Features and Benefits
- Corning® single-use spatulas are made of polystyrene and are color concentrate blends.
- Tapered blade/spoon with 247 mm length.
- Visually Inspected for accurate labeling, product configuration, seal and barrier integrity.
Legal Information
Corning is a registered trademark of Corning, Inc.
1 of 1
本產品 | |||
|---|---|---|---|
| description Tapered blade/spoon | description Round end/spoon | description flat end/spoon | description V-scoop/spoon |
| L 247 mm | L 248 mm | L 250 mm | L 250 mm |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| manufacturer/tradename Corning 3003 | manufacturer/tradename Corning 3005 | manufacturer/tradename Corning 3007 | manufacturer/tradename Corning 3006 |
Still not finding the right product?
Explore all of our products under Corning®一次性刮刀