AXYSCT150GS
1 of 10
Sigma-Aldrich
434R-1
ERG (EP111) Rabbit Monoclonal Primary Antibody
06-570
抗磷酸组蛋白H3(Ser10)抗体,有丝分裂标记
AXYSCT150OS
螺纹盖管和带 O 型圈的盖,圆锥形底部
C5992
Anti-Cytokeratin, pan antibody, Mouse monoclonal
MAB1612
Anti-Cytokeratin Epithelial Antibody, clone AE1
Millipore
1.17367
MAS-100 Atmos® Tri-Clamp (34 mm)
912719
(S)-BIDIME
Supelco
901922
Reducer (PE)
HS10060
带O型圈盖的螺口管
MTOX1301
Canine MDR1 Knockout MDCKII Control Cells
green screw top cap
sterile
RCF 20,000 × g
pack of 500 tubescase of 8 packs
Axygen IncCorning
1.5 mL
尋找類似的產品? 前往 產品比較指南
样品处理
輸入產品批次/批號來搜索 分析證明 (COA)。在產品’s標籤上找到批次和批號,寫有 ‘Lot’或‘Batch’.。
批次/批編號