文件
AXYSCT050SSYS
Self standing screw cap tubes and attached caps with O-rings
capacity 0.5 mL, yellow, sterile
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41121800
NACRES:
NB.11
材料
screw top cap
yellow
無菌
sterile
特點
RCF 20,000 × g
包裝
pack of 500 tubes
case of 8 packs
製造商/商標名
Axygen Inc
Corning
容量
0.5 mL