文件
21343
带端头的 Hamilton® 活塞组件,用于 1000、1700 和 1800 系列
for use with Hamilton 1701 (LT, N, TLL, RN), pkg of 1 ea
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41122001
材料
PTFE plunger tip
尺寸
10 μL
描述
1701, all
特點
PTFE tip
包裝
pkg of 1 ea
製造商/商標名
Hamilton 13205
進樣針體積
10 μL
相容性
for use with Hamilton 1701 (LT, N, TLL, RN)
尋找類似的產品? 前往 產品比較指南
一般說明
Hamilton® plunger assemblies with tips for series 1000, 1700, and 1800 are accessories for Hamilton syringes and these plungers do not fit sample lock syringes.
用于维修 Hamilton 1000、1700 和 1800 系列注射器。
法律資訊
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
分析證明 (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact 客戶支援