跳轉至內容
Merck
全部照片(1)

重要文件

364200

Sigma-Aldrich

GM 1489

同義詞:

GM 1489, N-[(2R)-2-(Carboxymethyl)-4-methylpentanoyl]-L-tryptophan-(S)-methyl-benzylamide

登入查看組織和合約定價


About This Item

經驗公式(希爾表示法):
C27H33N3O4
CAS號碼:
分子量::
463.57
分類程式碼代碼:
12352200

化驗

≥95% (HPLC)

品質等級

形狀

solid

製造商/商標名

Calbiochem®

儲存條件

OK to freeze

顏色

off-white

溶解度

DMSO: 25 mg/mL

運輸包裝

ambient

儲存溫度

−20°C

InChI

1S/C27H33N3O4/c1-17(2)13-20(15-25(31)32)26(33)30-24(14-21-16-28-23-12-8-7-11-22(21)23)27(34)29-18(3)19-9-5-4-6-10-19/h4-12,16-18,20,24,28H,13-15H2,1-3H3,(H,29,34)(H,30,33)(H,31,32)/t18-,20+,24-/m0/s1

InChI 密鑰

BLPWNGOQQLJQOL-NRYAXDJKSA-N

一般說明

This product has been discontinued.



A potent broad-range inhibitor of matrix metalloproteinases (MMPs). Inhibits MMPs in vitro (Ki = 0.2 nM for MMP-1; Ki = 500 nM for MMP-2; Ki = 20 µM for MMP-3; Ki = 100 nM for MMP-8; and Ki = 100 nM for MMP-9). Also reduces the inflammatory and the hyperproliferative responses that occur following topical phorbol ester application.

A potent broad-spectrum inhibitor of matrix metalloproteinases (MMPs). Inhibits MMPs in vitro [Ki = 0.2 nM for interstitial collagenase (MMP-1); Ki = 500 nM for gelatinase A (MMP-2); Ki = 20 µM for stromelysin (MMP-3); Ki = 100 nM for neutrophil collagenase (MMP-8); Ki = 100 nM for gelatinase B (MMP-9)]. Also reduces the inflammatory and the hyperproliferative responses that occur following topical phorbol ester application.

生化/生理作用

Cell permeable: no
Primary Target
MMP-1
Product does not compete with ATP.
Reversible: no
Target Ki: 0.2 nM for MMP-1; 500 nM for MMP-2; 20 µM for MMP-3; 100 nM for MMP-8; 100 nM for MMP-9

警告

Toxicity: Standard Handling (A)

重構

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

其他說明

Sato, N., et al. 2003. J. Natl. Cancer Inst.95, 327.
Holleran, W.M., et al. 1997. Arch. Dermatol. Res. 289, 138.

法律資訊

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

儲存類別代碼

10-13 - German Storage Class 10 to 13


分析證明 (COA)

輸入產品批次/批號來搜索 分析證明 (COA)。在產品’s標籤上找到批次和批號,寫有 ‘Lot’或‘Batch’.。

已經擁有該產品?

您可以在文件庫中找到最近購買的產品相關文件。

存取文件庫

我們的科學家團隊在所有研究領域都有豐富的經驗,包括生命科學、材料科學、化學合成、色譜、分析等.

聯絡技術服務