重要文件
Z341649
3M 6000/7000 series filters
A1 Organic Vapour filter, pkg of 2 ea
登入查看組織和合約定價
About This Item
分類程式碼代碼:
46182002
NACRES:
NB.35
描述
Protects against organic vapours with b.p. >65°C up to 10 × OEL* or 1000ppm*, EN141
包裝
pkg of 2 ea
製造商/商標名
3M 6051
存貨情形
not available in USA and Canada
尋找類似的產品? 前往 產品比較指南
一般說明
Gas and vapour filters
Two filters required per respirator.
特點和優勢
These filters utilise 3M′s advanced filter technology which minimises breathing resistance for greater comfort to the user. Select gas and vapour filters, particulate filters according to protection required. All filters fit both 6000 and 7000 series respirators by bayonet connection. Two filters required per respirator.
警告
All these filters should only be used for protection against hazards with good warning properties, e.g. smell or taste, and providing the concentrations of hazardous substances are not immediately dangerous to life or health (IDLH). *Where concentrations are given in terms of the OEL or ppm, the filter should only be used for protection against concentrations below the lower value.
法律資訊
3M is a trademark of 3M Company
從最近期的版本中選擇一個:
分析證明 (COA)