材料
stainless steel (with wire gauze)
製造商/商標名
Retsch
存貨情形
not available in USA and Canada
直徑 × 高度
200 mm × 50 mm
粒徑
2.8 mm
尋找類似的產品? 前往 產品比較指南
一般說明
For mechanical or manual, wet or dry sieving. Quintuple tested and provided with Quality Certificate DIN ISO 3310/1.