重要文件
Z301094
Quickfit® expansion adapter
top female joint: ST/NS 19/26, bottom male joint: ST/NS 14/23
同義詞:
XA21
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41121815
NACRES:
NB.35
接合處
female joint: ST/NS top19/26
male joint: ST/NS bottom14/23
製造商/商標名
Quickfit XA 21
存貨情形
not available in USA and Canada
not available in USA and Canada
尋找類似的產品? 前往 產品比較指南
一般說明
These adapters connect items with larger cone to a piece of glassware with a smaller size socket.
法律資訊
Quickfit is a registered trademark of SciLabware Limited
從最近期的版本中選擇一個:
分析證明 (COA)