跳轉至內容
Merck
全部照片(1)

重要文件

RNI00185

Sigma-Aldrich

(t-Bu)2PMe-Pd-G3

同義詞:

Methanesulfonato (di-tert-butyl) methylphosphino (2′-amino-1,1′-biphenyl-2-yl) palladium(II)

登入查看組織和合約定價


About This Item

經驗公式(希爾表示法):
C22H34NO3PPdS
CAS號碼:
分子量::
529.97
MDL號碼:
分類程式碼代碼:
12352200
PubChem物質ID:

特點

generation 3

反應適用性

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

官能基

phosphine

SMILES 字串

CP(C(C)(C)C)C(C)(C)C.NC1=C(C2=C([Pd]OS(C)(=O)=O)C=CC=C2)C=CC=C1

InChI

1S/C12H10N.C9H21P.CH4O3S.Pd/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-8(2,3)10(7)9(4,5)6;1-5(2,3)4;/h1-6,8-9H,13H2;1-7H3;1H3,(H,2,3,4);/q;;;+1/p-1

InChI 密鑰

SSBARXQHJYOPIL-UHFFFAOYSA-M

尋找類似的產品? 前往 產品比較指南

其他說明

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

儲存類別代碼

11 - Combustible Solids

水污染物質分類(WGK)

WGK 3

閃點(°F)

Not applicable

閃點(°C)

Not applicable


從最近期的版本中選擇一個:

分析證明 (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.

如果您需要協助,請聯絡 客戶支援

已經擁有該產品?

您可以在文件庫中找到最近購買的產品相關文件。

存取文件庫

我們的科學家團隊在所有研究領域都有豐富的經驗,包括生命科學、材料科學、化學合成、色譜、分析等.

聯絡技術服務