重要文件
BR65805
BRAND®干燥器
size 150 mm
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41104510
NACRES:
NB.43
描述
lid PC, base & desiccant tray PP
lid PC, base & desiccant tray PP
包裝
pack of 1 ea
製造商/商標名
BRAND 65805
尺寸
150 mm
尋找類似的產品? 前往 產品比較指南
相關類別
一般說明
Polycarbonate lid. PP base and desiccant tray. Venting stopper with non-return valve in lid. Seal between lid and base: polychlorobutadiene rubber (CK) O-ring, compressed by vacuum. Light-weight and easy to handle.
法律資訊
BRAND is a registered trademark of BRAND GMBH + CO KG
從最近期的版本中選擇一個:
分析證明 (COA)