重要文件
BAH379430000
SP Bel-Art® FEP Coated Steel Forceps
straight (fully coated)
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41122800
NACRES:
NB.43
材料
straight (fully coated)
包裝
pkg of 2 ea
製造商/商標名
Bel-Art H37943-0000
長度
11.4 cm (4½ in.) , long
尋找類似的產品? 前往 產品比較指南
相關類別
一般說明
These forceps are fully coated with brown PTFE FEP and comply with FDA regulations concerning direct food contact.
特點和優勢
  • Wide tips are perfect for lifting cover glasses
  • Available in two styles, bent or straight tip
法律資訊
Bel-Art is a registered trademark of SP Scienceware, a division of SP
從最近期的版本中選擇一個:
分析證明 (COA)