Z701033
Introduction to Macromolecular Crystallography
All Photos(1)
About This Item
ISBN-10:
0-471-25122-4
ISBN-13:
978-0-471-25122-4
UNSPSC Code:
55101509
publication info
A. McPherson, John Wiley & Sons, 2003, 237 pp., soft cover
General description
This book provides a comprehensive summary of the field of crystallography, fundamental theory of diffraction and properties of crystals, and applications in determining macromolecular structure. Without assuming a background in physics or math, book′s contents flow logically from basic principles to methods, such as those for solving phase problems, interpretation of Patterson maps, and the difference Fourier method. This introduction includes practical instruction on interpretation of data and methods for determining phases and also illustrates concepts throughout to clarify difficult ideas; integrates analogies to illuminate the text; equips the reader to employ the range of programs in crystallography.
Table of Contents
An Overview of Macromolecular Crystallography.
The Nature of Crystals: Symmetry and the Unit Cell.
Waves and Their Properties.
Diffraction from Points, Planes, Molecules, and Crystals.
Examination of Diffraction Patterns.
Solving the Phase Problem.
Interpreting Patterson Maps.
Electron Density Maps and Difference Fourier Maps.
Bibliography.
Index.
The Nature of Crystals: Symmetry and the Unit Cell.
Waves and Their Properties.
Diffraction from Points, Planes, Molecules, and Crystals.
Examination of Diffraction Patterns.
Solving the Phase Problem.
Interpreting Patterson Maps.
Electron Density Maps and Difference Fourier Maps.
Bibliography.
Index.
Choose from one of the most recent versions:
Certificates of Analysis (COA)