Skip to Content
Merck
All Photos(1)

Key Documents

SML2243

Sigma-Aldrich

BTSA1

≥98% (HPLC)

Synonym(s):

3-Phenyl-1-(4-phenyl-2-thiazolyl)-1H-pyrazole-4,5-dione 4-[2-(2-thiazolyl)hydrazone]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H14N6OS2
CAS Number:
Molecular Weight:
430.51
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

form

powder

color

red to dark red-brown

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=C1N(C2=NC(C3=CC=CC=C3)=CS2)N=C(C4=CC=CC=C4)/C1=N\NC5=NC=CS5

InChI key

KYVZVVOQWWCVPP-HKOYGPOVSA-N

Biochem/physiol Actions

BTSA1 is orally available, highly specific and potent BAX activator that binds to N-terminal activation site of Bax and induces conformational changes. BTSA1 potently induces apoptosis in leukemia cell lines and patient samples while sparing healthy cells.
orally available, highly specific and potent BAX activator that binds to N-terminal activation site of Bax

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denis E Reyna et al.
Cancer cell, 32(4), 490-505 (2017-10-11)
The BCL-2 family protein BAX is a central mediator of apoptosis. Overexpression of anti-apoptotic BCL-2 proteins contributes to tumor development and resistance to therapy by suppressing BAX and its activators. We report the discovery of BTSA1, a pharmacologically optimized BAX

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service