Skip to Content
Merck
All Photos(1)

Documents

P3656

Sigma-Aldrich

L-α-Phosphatidylethanolamine, dipalmitoyl, N-dansyl

2 mg/mL in chloroform/methanol (4:1), ~98%

Synonym(s):

1,2-Dipalmitoyl-sn-glycero-3-phospho-(N-dansyl)ethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H85N2O10PS
CAS Number:
Molecular Weight:
925.25
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

~98%

form

liquid

concentration

2 mg/mL in chloroform/methanol (4:1)

cation traces

Ca: ≤0.5 mol/mol

functional group

phospholipid

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OC[C@H](COP(O)(=O)OCCNS(=O)(=O)c1cccc2c(cccc12)N(C)C)OC(=O)CCCCCCCCCCCCCCC

InChI

1S/C49H85N2O10PS/c1-5-7-9-11-13-15-17-19-21-23-25-27-29-37-48(52)58-41-43(61-49(53)38-30-28-26-24-22-20-18-16-14-12-10-8-6-2)42-60-62(54,55)59-40-39-50-63(56,57)47-36-32-33-44-45(47)34-31-35-46(44)51(3)4/h31-36,43,50H,5-30,37-42H2,1-4H3,(H,54,55)/t43-/m1/s1

InChI key

QEYOOCBITXGEAT-VZUYHUTRSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D L Bratton et al.
Biochimica et biophysica acta, 943(2), 211-219 (1988-08-18)
Recent studies localizing the inflammatory mediator, platelet activating factor (PAF, 1-O-alkyl-2-acetyl-sn-glycero-3-phosphocholine), to the membranes of stimulated neutrophils, raise the possibility that PAF may, in addition to its activities as a mediator, alter the physical properties of membranes. This, and the
Guillaume Drin et al.
Biochimica et biophysica acta, 1559(2), 160-170 (2002-02-21)
The protegrin PG-1, belonging to the family of beta-stranded antimicrobial peptides, exerts its activity by forming pores in the target biological membranes. Linear analogues derived from PG-1 do not form pores in the phospholipid membranes and have been used successfully

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service