Skip to Content
Merck
All Photos(1)

Key Documents

I9281

Sigma-Aldrich

5′-Iodoresiniferatoxin

solid

Synonym(s):

6,7-Deepoxy-6,7-didehydro-5-deoxy-21-dephenyl-21-(phenylmethyl)-daphnetoxin,20-(4-hydroxy-5-iodo-3-methoxybenzeneacetate), I-RTX

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H39IO9
Molecular Weight:
754.60
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

color

white

solubility

DMSO: soluble 10 mg/mL

storage temp.

−20°C

SMILES string

COc1cc(CC(=O)OCC2=CC3C4OC6(Cc5ccccc5)O[C@]4(C[C@@H](C)[C@]3(O6)C7C=C(C)C(=O)[C@@]7(O)C2)C(C)=C)cc(I)c1O

InChI

1S/C37H39IO9/c1-20(2)35-16-22(4)37-26(33(35)45-36(46-35,47-37)18-23-9-7-6-8-10-23)12-25(17-34(42)29(37)11-21(3)32(34)41)19-44-30(39)15-24-13-27(38)31(40)28(14-24)43-5/h6-14,22,26,29,33,40,42H,1,15-19H2,2-5H3/t22-,26+,29-,33-,34-,35-,36?,37-/m1/s1

InChI key

TZUJORCXGLGWDV-RYACRXIZSA-N

Biochem/physiol Actions

Vanilloid receptor antagonist.

Legal Information

Sold in France, Germany, Great Britain and Switzerland under a patent license, restricted to research use only, from Procyon Pharmaceuticals, Inc.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nirlipta Swain et al.
Frontiers in cell and developmental biology, 10, 867057-867057 (2022-10-11)
Bi-directional crosstalk between Ca2+ signaling and ROS modulates physiological processes as a part of a regulatory circuit including sperm function. The role of transient receptor potential vanilloid 1 (TRPV1) in this regard cannot be undermined. This is the first report

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service