Skip to Content
Merck
All Photos(2)

Documents

GBL622101

Grace Bio-Labs CoverWell perfusion chambers

1 chamber, port diam. 1.5 mm, chamber diam. × thickness 20 mm × 0.9 mm

Synonym(s):

leak proof enclosure for slides, microscope stick on multiwell chamber, reusable press to seal multiwell chamber, water tight multiple well chamber

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122600
NACRES:
NB.22

material

clear polycarbonate backing
red silicon wells

feature

wells 1

packaging

pack of 40 ea

manufacturer/tradename

Grace Bio-Labs 622101

chamber diam. × thickness

20 mm × 0.9 mm

port diam.

1.5 mm

wells

1

Looking for similar products? Visit Product Comparison Guide

General description

CoverWell perfusion chambers press-to-seal covers form water-tight, multiwell cell incubation or cytochemistry chambers when pressed to coverslips or microscope slides. Reagents can be quickly added and removed through dual access ports without disturbing or cross-contaminating specimens. Chambers are available in five chamber configurations allowing up to eight different specimens or analytical tests on one coverglass.

CoverWell perfusion chambers form removable, water-tight multiwell enclosures by pressing to coverglass or microscope slides. Chambers are made of medical-grade silicone bonded to clear, UV-transparent plastic, and are removable for permanent mounting. The thin polycarbonate cover displays excellent transparency. The perfusion chambers are ideal for live cell imaging on upright or inverted microscopes at high magnification.

Legal Information

CoverWell is a trademark of Grace Bio-Labs, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service