Skip to Content
Merck
All Photos(1)

Key Documents

F9517

Sigma-Aldrich

(+)-Fenfluramine hydrochloride

Synonym(s):

(+)-N-Ethyl-α-methyl-m-[trifluoromethyl]phenethylamine

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352200

drug control

USDEA Schedule IV

Biochem/physiol Actions

Neurotoxic on prolonged administration or at high dosage; releases serotonin from axon terminals by a nonexocytotic mechanism; anorectic

Other Notes

Active isomer

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service