Skip to Content
Merck
All Photos(1)

Documents

E9134

Sigma-Aldrich

[3,5-diI-Tyr1, D-Ala2, N-Me-Phe4, Gly5-ol]-Enkephalin

≥97% (HPLC)

Synonym(s):

3,5-Diiodo-Tyr-D-Ala-Gly-N-Methyl-Phe-Gly-ol, [3,5-Diodo-Tyr1]-DAGO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H33I2N5O6
CAS Number:
Molecular Weight:
765.38
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CC(NC(=O)C(N)Cc1cc(I)c(O)c(I)c1)C(=O)NCC(=O)N(C)C(Cc2ccccc2)C(=O)NCCO

InChI

1S/C26H33I2N5O6/c1-15(32-25(38)20(29)12-17-10-18(27)23(36)19(28)11-17)24(37)31-14-22(35)33(2)21(26(39)30-8-9-34)13-16-6-4-3-5-7-16/h3-7,10-11,15,20-21,34,36H,8-9,12-14,29H2,1-2H3,(H,30,39)(H,31,37)(H,32,38)

InChI key

HVAFERGAOGMJBF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Selective μ opioid receptor agonist that can be tritiated for receptor binding studies.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service