Skip to Content
Merck
All Photos(1)

Documents

C5351

Sigma-Aldrich

Anti-Cyclin H antibody produced in rabbit

whole antiserum, aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

form

aqueous glycerol solution

mol wt

antigen 36 kDa

species reactivity

human

technique(s)

microarray: suitable
western blot: 1:500 using human HeLa cell nuclear extract

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CCNH(902)

Immunogen

recombinant human cyclin H, expressed in Escherichia coli.

Physical form

Solution of whole antiserum containing 30% glycerol and 0.014% thimerosal.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Rozek et al.
Polish journal of veterinary sciences, 16(4), 663-669 (2013-01-01)
Changes in the level of cellular proteins in cells inoculated with equine influenza virus H7N7 and H3N8 were studied with microarray technique. H3N8 induced pro-apoptotic proteins while H7N7 induced both pro- as well as anti-apoptotic factors. The higher level of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service