Skip to Content
Merck
All Photos(2)

Key Documents

AV41640

Sigma-Aldrich

Anti-SLC22A1 (AB2) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-1-Oct, Anti-HOCT1, Anti-Oct1_cds, Anti-Solute carrier family 22 (organic cation transporter), member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

56 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SLC22A1(6580)

General description

Solute carrier family 22 (organic cation transporter), member 1 (SLC22A1, 1-Oct, HOCT1, Oct1-cds) is a transporter of organic cations (OCT) that in addition to endogenous cations include various organic cationic toxins and drugs. Examples of drugs transported by OCT1 include metformin, amantadine, pramipexole, and, possibly, levodopa.

Specificity

Anti-SLC22A1 (AB2) polyclonal antibody reacts with human solute carrier family 22 (organic cation transporter), member 1.

Immunogen

Synthetic peptide directed towards the C terminal region of human SLC22A1

Application

Anti-SLC22A1 (AB2) polyclonal antibody is used to tag solute carrier family 22 (organic cation transporter), member 1 protein for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of solute carrier family 22 (organic cation transporter), member 1 in the transport of small organic cations including a variety of important drugs.

Biochem/physiol Actions

Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. The protein contains twelve putative transmembrane domains and is a plasma integral membrane protein.Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. This gene is one of three similar cation transporter genes located in a cluster on chromosome 6. The encoded protein contains twelve putative transmembrane domains and is a plasma integral membrane protein. Two transcript variants encoding two different isoforms have been found for this gene, but only the longer variant encodes a functional transporter.

Sequence

Synthetic peptide located within the following region: IAIQMICLVNAELYPTFVSGVGPACRGSDATSSRDQGGRFARDHEGRREP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service