All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C15H18N2O6S
CAS Number:
Molecular Weight:
354.38
MDL number:
UNSPSC Code:
12352201
suitability
suitable for preparation of neoglycoproteins
storage temp.
2-8°C
SMILES string
CC(=O)NC1C(O)C(O)C(CO)OC1Oc2ccc(cc2)N=C=S
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
M Monsigny et al.
Biology of the cell, 51(2), 187-196 (1984-01-01)
The presence and the sugar specificity of membrane lectins on the cell surface of mouse L1210 leukemia cells were demonstrated by using various neoglycoproteins (glycosylated serum albumin) substituted with fluorescein or methotrexate. Neoglycoproteins were prepared by reaction of glycosidophenylisothiocyanates with
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service