Skip to Content
Merck
All Photos(1)

Key Documents

A4911

Supelco

3-Acetamidophenol

Synonym(s):

3′-Hydroxyacetanilide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHC6H4OH
CAS Number:
Molecular Weight:
151.16
Beilstein:
907998
EC Number:
MDL number:
UNSPSC Code:
41116107
Pricing and availability is not currently available.

form

powder

color

yellow to gray

mp

145-148 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=O)Nc1cccc(O)c1

InChI

1S/C8H9NO2/c1-6(10)9-7-3-2-4-8(11)5-7/h2-5,11H,1H3,(H,9,10)

InChI key

QLNWXBAGRTUKKI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service