Skip to Content
Merck
All Photos(1)

Key Documents

BCR046

Benzo[b]chrysene

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H14
CAS Number:
Molecular Weight:
278.35
Beilstein:
2051199
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

c1ccc2cc3c(ccc4c5ccccc5ccc34)cc2c1

InChI

1S/C22H14/c1-2-7-17-14-22-18(13-16(17)6-1)10-12-20-19-8-4-3-5-15(19)9-11-21(20)22/h1-14H

InChI key

YYGRIGYJXSQDQB-UHFFFAOYSA-N

Analysis Note

For more information please see:
BCR046

Legal Information

BCR is a registered trademark of European Commission

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amir Mehravar et al.
Analytical methods : advancing methods and applications, 13(2), 242-249 (2020-12-23)
Chemometric and statistic approaches were applied to optimize and validate headspace-single drop microextraction-based deep eutectic solvents combined with gas chromatography-mass spectrometry. The proposed technique was used for the pre-concentration, separation, and detection of polyaromatic hydrocarbons in aqueous samples. The efficacy
Amir Mehravar et al.
Journal of chromatography. A, 1632, 461618-461618 (2020-10-21)
An improved deep eutectic solvent (DES)-based headspace single-drop microextraction procedure has been developed as a green procedure for gas chromatography-mass spectrometric analysis of polycyclic aromatic hydrocarbons (PAHs) in aqueous samples. The stability of the micro-drop was significantly improved using a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service