Skip to Content
Merck
All Photos(2)

Documents

MABT828

Sigma-Aldrich

Anti-Myoferlin Antibody, clone 159

culture supernatant, clone 159, from mouse

Synonym(s):

Myoferlin, Fer-1-like protein 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

159, monoclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... MYOF(26509)

General description

Myoferlin (UniProt Q9NZM1; also known as Fer-1-like protein 3) is encoded by the MYOF (also known as FER1L3, KIAA1207) gene (Gene ID 26509) in human. Myoferlin is a calcium/phospholipid-binding protein that is expressed in myoblast and endothelial cells and in cardiac and skeletal muscles. Low levels of myoferlin are seen in lung, and very low levels in kidney, placenta, and brain. Myoferlin plays a role in the plasmalemma repair mechanism of endothelial cells that permits rapid resealing of membranes disrupted by mechanical stress. Myoferlin contains C2 domains that participate in calcium-mediated membrane fusion events. It is also involved in endocytic recycling and has been implicated in VEGF signaling by regulating the levels of the receptor KDR. Myoferlin is overexpressed in breast cancer cells and its depletion blocks EGF-induced cell migration and epithelial to mesenchymal transition and reduces invasion through extracellular matrix (ECM). Ref.: Turtoi, A., et al. (2013). Cancer Res. 73, 5438-5448.

Specificity

Antibody blocking with the immunogen peptide prior to Western blotting abolished the target band detection (Jaiswal, J.K., et al. (2007). Traffic. 8(1):77-88). Epitope is absent from human isoform 4 and mouse isoform 3, but present in all other human and mouse spliced isoforms reported by UniProt (Q9NZM1 and Q69ZN7).

Immunogen

KLH-conjugated linear peptide corresponding to a sequence within the region between the second and the third C2 domain of human myoferlin.

Application

Immunocytochemistry Analysis: The undiluted hybridoma culture supernatant from a representative lot detected myoferlin in 4% paraformaldehyde-fixed, 0.3% Triton X-100-permeabilized C2C12 murine myoblasts.

Western Blotting Analysis: A representative lot detected myoferlin in C2C12 murine myoblast lysate and human skin fibroblast lysate. Antibody blocking with the immunogen peptide prior to Western blotting abolished the target band detection (Jaiswal, J.K., et al. (2007). Traffic. 8(1):77-88).
Research Category
Cell Structure
This mouse monoclonal hybridoma culture supernatant, Anti-Myoferlin, clone 159, Cat. No. MABT828, is validated for use in Immunocytochemistry and Western Blotting for the detection of Myoferlin.

Quality

Evaluated by Western Blotting in C2C12 cell lysate.

Western Blotting Analysis: A 1:10 dilution of this hybridoma culture supernatant detected myoferlin isoforms in 10 µg of C2C12 murine myoblast lysate.

Target description

~233/207/103 kDa observed. 234.7/229.9/233.3/179.6/233.5/49.72/46.70 kDa (human isoform 1/2/3/5/6/7/8), 233.3/103.4/207.6 kDa (mouse isoform 1/2/4) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Mouse IgG2aκ hybridoma culture supernatant without azide.
Unpurified.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service