Skip to Content
Merck
All Photos(1)

Key Documents

MABC1144

Sigma-Aldrich

Anti-LMO-2 (Rhombotin-2) Antibody, clone 1A9-3B11

clone 1A9-3B11, from mouse

Synonym(s):

Rhombotin-2, Cysteine-rich protein TTG-2, LIM domain only protein 2, LMO-2, T-cell translocation protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
clone:
1A9-3B11, monoclonal
application:
IHC
species reactivity:
human
technique(s):
immunohistochemistry: suitable (paraffin)
citations:

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1A9-3B11, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... LMO2(4005)

General description

Rhombotin-2 (UniProt P25791; also known as Cysteine-rich protein TTG-2, LIM domain only protein 2, LMO-2, T-cell translocation protein 2) is encoded by the LMO2 (also known as RBTN2, RBTNL1, RHOM2, TTG2) gene (Gene ID 4005) in human. Rhombotin-2 is 158 amino acids, small cysteine-rich protein with multiple functions. A large portion of its structure is occupied by tandemly repeated cysteine-rich zinc-finger binding motifs, the LIM domains. However, it does not have any identifiable DNA-binding motifs. It is found predominantly in the nucleus or in the cytoplasm depending upon both the tissue and the cell type. For example, in kidney it is predominantly localized in the cytoplasm, but in hematopoietic cells it has nuclear localization. LMO2 is reported to be a proto-oncogene only in the context of T lymphocytes and plays a role in T-cell leukemogenesis. It also has distinct functions in erythropoiesis. It acts with TAL1/SCL to regulate red blood cell development and interacts with LDB1 to maintain erythroid precursors in an immature state. It is reported to interact with retinoblastoma-binding protein 2 (RBP2) and human Bex2 (brain expressed X-linked), both of which are shown to enhance its transcriptional activity. A chromosomal aberration involving LMO2 has been linked to T-cell acute lymphoblastic leukemia (T-ALL).
Ref.: Mao S., et al. (1997) Oncogene 14, 1531-1539.

Immunogen

GST-tagged recombinant human LMO-2 (Rhombotin-2).

Application

Detect LMO2 using this mouse monoclonal Anti-LMO-2 (Rhombotin-2) Antibody, clone 1A9-3B11, Cat. No. MABC1144, validated for use in Immunohistochemistry (Paraffin).
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected LMO-2 (Rhombotin-2) in human lymphoma and cerebellum tissue sections.

Clone 1A9-3B11 exhibits similar property as clone 1A9-1 subcloned from the same parental hybridoma culture (Natkunam, Y., et al. (2007). Blood. 109(4):1636-1642). Please refer to Natkunam, Y., et al. (2007) regarding Immunohistochemistry and Western Blotting applications using the related subclone 1A9-1.
Research Category
Apoptosis & Cancer

Quality

Evaluated by Immunohistochemistry in human kidney tissue sections.

Immunohistochemistry Analysis: A 1:1,000 dilution of this antibody detected LMO-2 (Rhombotin-2) in human kidney tissue section.

Target description

18.36/25.04 kDa (human isoform 1/3) calculated. ~24 kDa reported (Natkunam, Y., et al. (2007). Blood. 109(4):1636-1642).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service