Skip to Content
Merck
All Photos(1)

Documents

IPAKGARA1

Milli-Q

IPAK Gard® Pretreatment Pack

This product is replaced with IPAKGARD1. Search to Explore!

Synonym(s):

IPAK Gard® 3-5 pretreatment pack

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212
eCl@ss:
33050190
NACRES:
NB.85

Quality Level

feature

removal of colloids, particles and free chlorine

packaging

pkg of 1 unit

manufacturer/tradename

IPAK Gard®

H

39.5 cm (15.5 in.)

pore size

5 μm pore size

compatibility

for use with Milli-Q® EQ 7008
for use with Milli-Q® EQ 7016
for use with Milli-Q® IQ 7003
for use with Milli-Q® IQ 7005

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

IPAK Gard® 3-5 pretreatment pack provides optimal protection of the reverse osmosis (RO) membrane, improving your system’s performance and extending its lifetime. For use with Milli-Q® IQ 7003/05, Milli-Q® IX 7003/05 and Milli-Q® EQ 7008/16 water purification systems.

Features and Benefits

  • Pleated filter and carbon block components provide high-efficiency removal of colloids, particles and free chlorine from potable water to ensure reverse osmosis (RO) membrane performance over years.
  • 63% of the parts suppliers are ISO 14001 environmental management system-certified.

Legal Information

IPAK GARD is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service