Skip to Content
Merck
All Photos(1)

Key Documents

70623

Sigma-Aldrich

Tuner (DE3) Competent Cells - Novagen®

Escherichia coli, rod shaped

Synonym(s):

Novagen Competent Cells, Tuner Competent Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106202
Pricing and availability is not currently available.

Product Name

Tuner (DE3) Competent Cells - Novagen®, Tuner host strains are lacZY deletion mutants of BL21, which enable adjustable levels of protein expression throughout all cells in a culture.

biological source

Escherichia coli

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

growth mode

adherent or suspension

morphology

rod shaped

technique(s)

microbiological culture: suitable

cell transformation

transformation efficiency: >2×106 cfu/μg

shipped in

dry ice
wet ice

storage temp.

−70°C

General description

Genotype: FompT hsdSB (rB mB) gal dcm lacY1(DE3)





This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges us to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.
Tuner strains are lacZY deletion mutants of BL21 and enable adjustable levels of protein expression throughout all cells in a culture. The lac permease (lacY) mutation allows uniform entry of IPTG into all cells in the population, which produces a concentration-dependent, homogeneous level of induction. By adjusting the concentration of IPTG, expression can be regulated from very low levels up to the robust, fully induced levels commonly associated with pET vectors. Lower-level expression may enhance the solubility and activity of difficult target proteins.

DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
Tuner host strains are lacZY deletion mutants of BL21, which enable adjustable levels of protein expression throughout all cells in a culture.

Components

0.4 ml1 mlComponent

•2 × 0.2 ml5 × 0.2 mlTuner(DE3) Competent Cells

•2 × 2 ml4 × 2 mlSOC Medium

•2 ng2 ngTest Plasmid

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamed A Abouelkhair et al.
Virulence, 9(1), 1050-1061 (2018-07-28)
Staphylococcus pseudintermedius is an opportunistic pathogen that is the major cause of pyoderma affecting dogs. Conventional antimicrobial treatment for infections caused by this organism have failed in recent years due to widespread resistance and alternative treatment strategies are a high
Zhitai Hao et al.
Molecular cell, 81(2), 281-292 (2020-12-10)
Rho is a general transcription termination factor playing essential roles in RNA polymerase (RNAP) recycling, gene regulation, and genomic stability in most bacteria. Traditional models of transcription termination postulate that hexameric Rho loads onto RNA prior to contacting RNAP and
Dieter Eibl et al.
Chimia, 75(3), 215-218 (2021-03-27)
When the SARS-CoV-2 pandemic started,[1] science came to the immediate attention of the broad public. People and politicians were hanging on every word of medical doctors, virologists, molecular biologists, data scientists and many others in the hope of finding other
Erika Zangelmi et al.
iScience, 26(11), 108108-108108 (2023-10-25)
Phosphonates-compounds containing a direct C-P bond-represent an important source of phosphorus in some environments. The most common natural phosphonate is 2-aminoethylphosphonate (AEP). Many bacteria can break AEP down through specialized "hydrolytic" pathways, which start with the conversion of AEP into
Preparation of Cas9 Ribonucleoproteins for Genome Editing.
Lin, et al.
Bio-protocol, 12, e4420-e4420 (2022)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service