Skip to Content
Merck
All Photos(1)

Documents

69171-M

Sigma-Aldrich

HSV-Tag® Monoclonal Antibody

Novagen®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

storage temp.

−20°C

target post-translational modification

unmodified

General description

The HSV•Tag Antibody is a mouse monoclonal antibody (IgG1) that has high specificity and affinity for an 11-amino acid peptide derived from Herpes Simplex Virus (HSV) glycoprotein D. The peptide is encoded by the HSV•Tag sequence in appropriate expression vectors and by the Positive Control 36mer in the Novagen NovaTope System. The purified antibody produces a strong signal at a 1:10,000 dilution on Western blots (Sakai 1996), dot blots, and colony lifts when used with the Novagen Anti-Mouse IgG AP Conjugate.

Sensitive, highly specific detection of HSV•Tag fusion proteins

Components

•: 40 µg: HSV•Tag Monoclonal Antibody (1 mg/ml solution)


•: : Positive Control Extract containing 31.1 kDa target protein

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Other Notes

Sakai, J., et al. 1996. Cell85, 1037.

Legal Information

HSV-TAG is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service