Skip to Content
Merck
All Photos(1)

Key Documents

6.08101

Sigma-Aldrich

Tetrahydrofuran

for chromatography

Synonym(s):

Tetrahydrofuran, THF, Butylene oxide, Oxolane, Tetramethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
Beilstein:
102391
MDL number:
UNSPSC Code:
12352005

grade

for chromatography

Quality Level

vapor density

2.5 (vs air)

vapor pressure

114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

autoignition temp.

610 °F

expl. lim.

1.8-11.8 %

refractive index

n20/D 1.407 (lit.)

bp

65-67 °C (lit.)

mp

−108 °C (lit.)

density

0.889 g/mL at 25 °C (lit.)

suitability

suitable for chromatography

SMILES string

C1CCOC1

InChI

1S/C4H8O/c1-2-4-5-3-1/h1-4H2

InChI key

WYURNTSHIVDZCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrahydrofuran(THF) is commonly used as a solvent in liquid chromatography. THF is widelyutilized as a solvent in non-aqueous size-exclusion chromatography (SEC),because it is a good solvent for various polymer types.

Application

Tetrahydrofuran can be used as a mobile-phase component in the:
  • Analysis of tocopherols.
  • HPLC analysis of self-associated cationicpolymer (NREP).

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-6.2 °F - closed cup

Flash Point(C)

-21.2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The thermodynamic properties of tetrahydrofuran-water association and its influence on retention in liquid chromatography
Scott RPW
Journal of Liquid Chromatography and Related Technologies, 23(10), 3083-3093 (2000)
Designing a self-associated cationic polymer for enhanced compatibility, palatability, and gastric release of cefuroxime axetil
Menjoge AR, et al.
Biomacromolecules, 8(2), 532-542 (2007)
The drop-in-drop encapsulation in chitosan and sodium alginate as a method of prolonging the quality of linseed oil
Mania S, et al.
Polymer, 10(12), 1355-1355 (2018)
Organic solvent modifier and temperature effects in non-aqueous size-exclusion chromatography on reversed-phase columns
Caltabiano AM, et al.
Journal of Chromatography A, 1531, 83-103 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service