219489
Ceramide Kinase Inhibitor, K1
Synonym(s):
Ceramide Kinase Inhibitor, K1, F-12509A Cyclic Product K1
About This Item
Assay
≥95% (mixture of enantiomers, NMR)
Quality Level
form
powder
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
yellow
solubility
ethanol: 20 mg/mL
shipped in
wet ice
storage temp.
−20°C
General description
This product has been discontinued.
A cell-permeable tetracyclic quinone compound that acts as a specific, reversible, and non-competitive inhibitor of CerK (ceramide kinase) activity (IC50 = ~5 µM) with little effect against SPHK1/2 or DGKγ (IC50 >100 µM). Shown to reduce cellular C1P (ceramide-1-phosphate) synthesis by 40% at ≥ 20 µM in a rat basophilic leukemia cell line RBL-2H3 and block CerK-mediated degranulation in both RBL-2H3 and murine BMMC (bone marrow-derived mast cells) in a dose-dependent manner. Exhibits no cytotoxic effect against RBL-2H3 even at concentrations as high as 100 µM.
Packaging
Warning
Reconstitution
Other Notes
Kumada, H., et al. 2007. Biosci. Biotechnol. Biochem.71, 2581
Kim, J.W., et al. 2005. Biochim. Biophys. Acta1738, 82.
Legal Information
Storage Class Code
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service