Skip to Content
Merck
All Photos(5)

Key Documents

15-109

Sigma-Aldrich

SAPK Pathway Explorer Antibody Minipack

Upstate®

Synonym(s):

activating transcription factor 2, activating transcription factor 2 splice variant ATF2-var2, cAMP response element-binding protein CRE-BP1 , cAMP responsive element binding protein 2, mitogen-activated protein kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.77
application:
citations:

manufacturer/tradename

Upstate®

Quality Level

UniProt accession no.

shipped in

dry ice

Gene Information

human ... MAP2K6(5608)

General description

Pathway Explorer Antibody MiniPack:
Each Pathway Explorer Antibody Minipack contains three related antibodies as part of a signaling cascade or a combination of total and phosphorylated forms of key signaling targets. Each of the three antibodies are 30% the original pack size. Full size versions of each of the Pathway Explorer antibodies are available for sale individually under the same catalog number with the removal of “SP” off of each one (e.g. 05-591SP can be ordered as 05-591).

MKK6/SKK3:
MKK6 (MAP2K6, MEK6) is a dual kinase that phosphorylates both threonine and tyrosine residues in the TxY activation motif of p38 just as MEK1/2 does for Erk1/2. The p38 Mitogen-Activated Protein Kinase (MAPK) family is a subfamily of the JNK/SAPK family that have traditionally been associated with stress, immune response, and regulation of cel proliferation. They are activated by treatment of cells with cytokines and by exposure to environmental stress. The effects of these stimuli on p38 MAP Kinase are mediated by the MAP Kinase Kinases (MKKs) MKK3, MKK4, and MKK6. MKK6 appears to be involved in cell cycle, transcription, and apoptosis.

p38α:
The p38 family of kinases, also known as the SAPK2/3 Kinases, are a subfamily of JNK/SAPK family that have traditionally been associated with stress, immune response, and regulation of cell proliferation. The p38 MAPK family of Ser/Thr kinases, referred to as the stress-activated MAPK, is made up of 4 isoforms α,β, δ, and γ. They are regulated by stress-inducing signals such as osmotic stress, UV radiation, pro-inflammatory cytokines, and certain toxins such as anisomycin. Upon stimulation, they are dually phosphorylated on their common TGY motif resulting in their activation, which is Thr180/Tyr182 in human p38α. Once activated, p38 is able to phosphorylate a wide array of targets including numerous kinases such as MAPKAP-K2 and ATF-2. p38 has been implicated in many pathological conditions that include inflammation, rheumatoid arthritis, diabetes, cancer, and neurodegenerative diseases.

ATF2:
Activating transcription factor 2 (ATF2) is a member of the basic region leucine zipper (bZIP) family of proteins. Transcriptionally active ATF2 recognizes and binds to specific ATF/CRE motifs as homo- or heterodimers with other bZIP transcription factors of the ATF/CREB or AP-1 families. ATF2 is regulated by phosphorylation on Thr69/71. Two major kinases known to phosphorylate these sites are the two Stress Activated Protein Kinases (SAPK) Jun and p38 that are mitogen activated protein kinases (MAPKs). Phosphorylation and dimerization are required for the full transcriptional activity of ATF2. Transcriptional regulation by ATF2 has been implicated in mediating many diverse cellular processes such as growth, differentiation, immune and stress responses, inflammatory cytokine signaling, and oncogenic transformation.

*See full size versions for corresponding references.

Application

Research Category
Signaling

Apoptosis & Cancer
Research Sub Category
MAP Kinases

Transcription Factors
This Antibody pack contains Anti-MKK6/SKK3 Antibody, Anti-phospho-p38α Antibody(Thr180/Tyr182), Anti-phospho-ATF2 Antibody(Thr69/71).

Packaging

Each vial is 30% the size of the parent catalog number

Components

07-417SP Anti-MKK6/SKK3

09-272SP Anti-phospho-p38α (Thr180/Tyr182)

05-891SP Anti-phospho-ATF2 (Thr69/71),
clone AW65

Physical form

3 individual tubes containing either Anti-MKK6/SKK3; Anti-phospho-p38α (Thr180/Tyr182): or Anti-phospho-ATF2 (Thr69/71),
clone AW65

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service