Skip to Content
Merck
All Photos(1)

Key Documents

14-733-M

Sigma-Aldrich

c-Kit (V654A) Protein, active, 10 µg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

biological source

human

Quality Level

mol wt

Mw 76.7 kDa

manufacturer/tradename

Chemicon®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

General description

N-terminal GST tagged, recombinant human cKit, amino acids 544 - end containing the V654A mutation. Co-ordinates encompass the cytoplasmic domain (544-976) according in Swiss Prot P10721. V654A is an acquired resistance mutation which has been found in patients with gastrointestinal stromal tumours (GISTs) undergoing treatment with Imatinib. The V654A mutation is strongly correlated with imatinib resistance and rapid progession of GISTs. (Chen LL et al., Cancer Research, (2004); 64:5913-5919).
recombinant protein expressed in Sf21 cells

Application

Research Category
Metabolism|Inflammation & Immunology
Research Sub Category
Obesity|Metabolic Disorders|Osteoporosis|Arthritis
V654A is an acquired resistance mutation found in patients with gastrointestinal stromal tumours (GISTs) undergoing treatment with Imatinib. The V654A mutation is strongly correlated with imatinib resistance & rapid progession of GISTs.

Biochem/physiol Actions

  • Protein Target: c-Kit
  • Target Sub-Family:TK

Quality

Routinely evaluated by phosphorylation of 250 μM GGMEDIYFEFMGGKKK.
Purity was assessed by SDS-PAGE and Coomassie blue staining using 3 μg of active cKit (V654A).

Physical form

Format: Purified using glutathione-agarose. Purity 99% by SDS-PAGE and Coomassie blue staining.

Storage and Stability

Stable for 6 months at -70 °C from date of shipment. For maximum recovery of product, centrifuge original vial prior to removing the cap.
Rapidly thaw the vial under cold water and immediately place on ice. Aliquot unused material into pre-chilled

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service