Skip to Content
Merck
All Photos(1)

Documents

1.46669

Millipore

Tryptic Soy Broth Ready-to-use Bottles

with Tween® 1 %, bottle capacity 500 mL, bottle filling volume 500 mL, closure type, blue screw cap and septum, box of 6 bottles

Synonym(s):

CASO, Casein Soybean Digest Broth with 0.1 % Polysorbate, SCDB, TSB, Tryptic Soy Broth with 1% Tween® 80

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106214

Agency

EP 2.6.1
JP 4.06
USP 71

Quality Level

sterility

sterile; autoclaved

form

liquid

feature

closure type blue screw cap and septum
ready-to-use

composition

with Tween®, 1%

packaging

box of 6 bottles

bottle capacity

500 mL

bottle filling volume

500 mL

pH

7.3±0.2

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

storage temp.

2-25°C

suitability

nonselective for

Looking for similar products? Visit Product Comparison Guide

General description

Tryptic Soy Broth with 1% Tween® 80 is a nutrient rich inhibitor free microbial media designed in compliance with regulatory bodies like European (EP), Japanese (JP) and United States Pharmacopeia (USP).
The peptones present come from casein and soymeal which provides an enhanced carbon source, which promotes the growth of the microbes present in the test sample.
Polysorbate 80 (Tween®) is a neutralizer to inactivate residues of disinfectants containing active agents such as Alcohol (70% ethanol or isopropyl alcohol), Aldehyde, Dichloroisocyanurate, Glucoprotamine, Peracetic acid, Phenols, Biguanides and quaternary ammonium compounds.

Application

Tryptic Soy Broth with 1% Tween® 80 is used for the isolation and cultivation of fastidious bacteria, yeast. TSB is suitable for sterility testing of substances, preparations, and products from pharmaceutical and food industry.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service