Skip to Content
Merck
All Photos(1)

Documents

1.03873

Millipore

Sabouraud Dextrose Agar

GranuCult® prime, with 2% Dextrose, according to ISO, granular, pkg of 500 g

Synonym(s):

SAB, SDA, Sabouraud 2% glucose agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Agency

according to ISO

Quality Level

form

granular

composition

with 2% Dextrose

packaging

pkg of 500 g

manufacturer/tradename

GranuCult® prime

storage condition

(Tightly closed. Dry)

technique(s)

microbiological culture: suitable

color

beige Dehydrated medium
clear to slightly opaque yellowish to brown Prepared medium

application(s)

food and beverages
pharmaceutical

suitability

nonselective for molds
nonselective for yeasts

Looking for similar products? Visit Product Comparison Guide

General description

Sabouraud Dextrose Agar contains peptones to provide nitrogen, vitamins, and amino acids. The relatively high concentration of glucose (dextrose), in addition to the low pH, promotes the growth and the formation of spores (Conidia and Sporangia) and pigments of yeasts and molds. The low pH further inhibits the growth of bacteria. Agar is the solidifying agent.

Application

Sabouraud Dextrose Agar is suitable for the cultivation of yeasts and molds from different materials and for usage as a reference medium during performance testing of culture media.

Features and Benefits

  • Safe and sustainable due to reduced risks associated with fine dust and toxic substance inhalation, resulting in a safer work environment.
  • Excellent wettability, solubility, and free flowing properties
  • Convenient, with minimal component separation and clumping, even under warm or humid conditions
  • High batch-to-batch reproducibility
  • Prolonged shelf life of up to five years
  • High number of test strains exceeding all regulatory demands
  • Granulation technology allowing many supplements to be included, with no need to add these separately

Storage and Stability

  • Store at +15 °C to +25 °C, dry and tightly closed.
  • Do not use clumped or discolored medium.
  • Protect from UV light (including sunlight).
  • For in vitro use only.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service